Brucin-sulfat Heptahydrat ACS

Lieferant: Thermo Scientific
Gefahr

36249.14
36249.14EA 114 EUR
36249.14
Brucin-sulfat Heptahydrat ACS
Brucine sulphate heptahydrate

CAS No.: 60583-39-3 Molecular Formula: (C23H26N2O4)2.H2SO4.7H2O Formula Weight: 1013.15 (887.03anhy) Physical Form: Powder Melting Point: 180° dec. UN No.: UN2811 MDL No.: MFCD00150159


Applications: Resolving agent

Formel: C₂₃H₂₆N₂O₄
Molecular Weight: 887,01 g/mol
Schmelzpunkt: 180 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013472
CAS-Nummer: 60583-39-3
EINECS: 225-432-9
UN: 2811
ADR: 6.1,I
Merck Index: 14,1455

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR