Zirkonium ≥99,8% Metall-Basis ausgeschlossen Hf, Klumpen 3-6 mm (0.1-0.2 in) (max. 4% Hf)
Lieferant: Thermo Fisher Scientific
36253.14
36253.22
36253.14EA
77.6
EUR
36253.14
36253.22
Zirkonium ≥99,8% Metall-Basis ausgeschlossen Hf, Klumpen 3-6 mm (0.1-0.2 in) (max. 4% Hf)
Zirkonium
Purity: 99.8%
CAS No.: 7440-67-7
Physical Form: For vacuum deposition
Odour: Odourless
MDL No.: MFCD00011303
Formel:
Zr Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011303 CAS-Nummer: 7440-67-7 EINECS: 231-176-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...