2-Hydroxy-2-propanesulphonic acid monosodium salt ≥97%

Lieferant: Thermo Fisher Scientific

36256.22 36256.36
36256.22EA 102 EUR
36256.22 36256.36
2-Hydroxy-2-propanesulphonic acid monosodium salt ≥97%
2-Hydroxy-2-propanesulphonic acid monosodium salt

Purity: 97% CAS No.: 540-92-1 Molecular Formula: C3H7NaO4S Formula Weight: 162.14 Physical Form: Crystalline MDL No.: MFCD00031395

Formel: C₃H₇NaO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD00031395
CAS-Nummer: 540-92-1
EINECS: 208-761-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR