Chrom(III)oxid ≥99,6% (Metall-Basis), verschmolzener Klumpen 3-6mm

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Chromgrün

36258.14 36258.22
36258.14EA 52.9 EUR
36258.14 36258.22
Chrom(III)oxid ≥99,6% (Metall-Basis), verschmolzener Klumpen 3-6mm
Chrom(III)oxid

For vacuum deposition


It can be used in selective oxidation. It is mainly used for smelting metal chromium and chromium carbide. It is used as coloring agent for color painting on enamelware and porcelain ware, imitation leather or building material. It is also used for making sun-proof coating, abrasive, green polishing compound and special printing ink for printing bank note. It is used as catalyst for organic synthetic chromium.


For vacuum deposition

Formel: Cr₂O₃
Siedepunkt: 3000 °C
Schmelzpunkt: 2435 °C
Dichte: 5,22
Lagertemperatur: Raumtemperatur
MDL: MFCD00010949
CAS-Nummer: 1308-38-9
EINECS: 215-160-9
Merck Index: 14,2234

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR