Praseodym(III,IV)oxid 99,9

Lieferant: Thermo Fisher Scientific
Achtung

36276.09 36276.18
36276.09EA 73.6 EUR
36276.09 36276.18
Praseodym(III,IV)oxid 99,9
Praseodym(III,IV)oxid

Purity: 99.9% CAS No.: 12037-29-5 Molecular Formula: Pr6O11 Formula Weight: 1021.43 Physical Form: 3-12mm Sintered Lump Melting Point: 2500° MDL No.: MFCD00011178

Formel: O₁₁Pr₆
Molecular Weight: 1021.44 g/mol
Schmelzpunkt: 2500 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011178
CAS-Nummer: 12037-29-5
EINECS: 234-857-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR