Samarium(III)oxid ≥99,9% (Metall-Basis)

Lieferant: Thermo Fisher Scientific

36278.09 36278.18
36278.09EA 108 EUR
36278.09 36278.18
Samarium(III)oxid ≥99,9% (Metall-Basis)
Samarium(III)oxid

Purity: 99.9% CAS No.: 12060-58-1 Molecular Formula: Sm2O3 Formula Weight: 348.70 Physical Form: 3-12mm Sintered Lump Refractive Index: 1.97 Melting Point: 2300° Merck Reference: 14,8347 MDL No.: MFCD00011237

Formel: O₃Sm₂
Schmelzpunkt: 2300 °C
Dichte: 7,68 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011237
CAS-Nummer: 12060-58-1
EINECS: 235-043-6
Merck Index: 14,8347

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR