Thulium(III)oxid ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
36279.04
36279.09
36279.04EA
152
EUR
36279.04
36279.09
Thulium(III)oxid ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Thulium(III)oxid
Purity: 99.9%
CAS No.: 12036-44-1
Molecular Formula: O3Tm2
Formula Weight: 385.87
Physical Form: 3-12mm Sintered Lump
Appearance: Pale green
Melting Point: 2425°
Merck Reference: 14,9394
MDL No.: MFCD00011285
Formel:
O₃Tm₂ Schmelzpunkt: 2425 °C Dichte: 8,6 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011285 CAS-Nummer: 12036-44-1 EINECS: 234-851-6 Merck Index: 14,9394 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...