Essigsäureanhydrid 97+% ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Acetanhydrid
36292.AP
36292.K3
36292.AE
36292.APEA
75.6
EUR
36292.AP
36292.K3
36292.AE
Essigsäureanhydrid 97+% ACS
Essigsäureanhydrid
Purity: 97+%
CAS No.: 108-24-7
Molecular Formula: C4H6O3
Formula Weight: 102.09
Flash Point: 49°(120°F)
Physical Form: Liquid
Density: 1.082
Refractive Index: 1.3900
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Pungent
Melting Point: -73°
Boiling Point: 138-140°
Beilstein Reference: 385737
Merck Reference: 14,56
UN No.: 1715
MDL No.: MFCD00008705
Formel:
(CH₃CO)₂O Molecular Weight: 102.09 g/mol Siedepunkt: 138…140 °C Schmelzpunkt: –73 °C Dichte: 1.082 Flammpunkt: 49 °C (120 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008705 CAS-Nummer: 108-24-7 EINECS: 203-564-8 UN: 1715 ADR: 8 II REACH: 01-2119486470-36 Merck Index: 14,56 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...