Dithalliumselenid ≥99,9% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Thallium(I) selenide

36296.09
36296.09EA 545 EUR
36296.09
Dithalliumselenid ≥99,9% (Metall-Basis)
Dithalliumselenid

Purity: 99.9% CAS No.: 15572-25-5 Molecular Formula: Tl2Se Formula Weight: 487.70 Physical Form: 10mm & Down Lump Density: 9.05 Melting Point: ca 340° Solubility: Insoluble in water, acids Merck Reference: 14,9267 UN No.: 1707 MDL No.: MFCD00049603

Formel: SeTl₂
Molecular Weight: 487.73 g/mol
Schmelzpunkt: ∼340 °C
Dichte: 9.05
Lagertemperatur: Raumtemperatur
MDL: MFCD00049603
CAS-Nummer: 15572-25-5
EINECS: 239-627-1
UN: 1707
ADR: 6.1 II
Merck Index: 14,9267

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR