Indium tin oxide ≥99,99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
36348.06
36348.14
36348.22
36348.06EA
82.6
EUR
36348.06
36348.14
36348.22
Indium tin oxide ≥99,99% (Metall-Basis)
Indium tin oxide
Purity: 99.99%
CAS No.: 50926-11-9
Molecular Formula: In2O3:SnO2; 90:10 wt%
Physical Form: -325+500 Mesh Powder
Odour: Odourless
MDL No.: MFCD00171662
Formel:
In₂O₃·(SnO₂)x Lagertemperatur: Raumtemperatur |
MDL:
MFCD00171662 CAS-Nummer: 50926-11-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...