Bis(tri-tert-butylphosphin)palladium(0) 98%

Lieferant: Thermo Fisher Scientific
Achtung

363501000 363500010
363501000.EA 57 EUR
363501000. 363500010.
Bis(tri-tert-butylphosphin)palladium(0) 98%
Bis(tri-tert-butylphosphin)palladium(0)
Formel: C₂₄H₅₄P₂Pd
Schmelzpunkt: 258…272 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03094580
CAS-Nummer: 53199-31-8

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to orange-brown Powder or crystals
Infrared spectrum Conforms
Assay ≥97.5 % (ex Pd)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR