Bariumfluorid ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
36380.14
36380.14EA
170
EUR
36380.14
Bariumfluorid ≥99,9% (Metall-Basis)
Barium fluoride
Purity: 99.9%
CAS No.: 7787-32-8
Molecular Formula: BaF2
Formula Weight: 175.36
Physical Form: 3-6mm Fused Lump
Density: 4.83
Refractive Index: 1.4741
Odour: Odourless
Melting Point: 1353°
Boiling Point: 2260°
Merck Reference: 14,974
UN No.: 1564
MDL No.: MFCD00003450
Formel:
BaF₂ Siedepunkt: 2260 °C Schmelzpunkt: 1353 °C Dichte: 4,83 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003450 CAS-Nummer: 7787-32-8 EINECS: 232-108-0 UN: 1564 ADR: 6.1,III Merck Index: 14,974 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...