Indium(III)oxid ≥99,999% (Metall-Basis)

Lieferant: Thermo Fisher Scientific

36386.09 36386.18
36386.09EA 131 EUR
36386.09 36386.18
Indium(III)oxid ≥99,999% (Metall-Basis)
Indium(III)oxid

Purity: 99.999% CAS No.: 1312-43-2 Molecular Formula: In2O3 Formula Weight: 277.64 Physical Form: 3-6mm Sintered Granules Density: 7.18 Melting Point: 850° volat. Merck Reference: 14,4952 MDL No.: MFCD00011060

Formel: In₂O₃
Schmelzpunkt: 850 °C
Dichte: 7,18
Lagertemperatur: Raumtemperatur
MDL: MFCD00011060
CAS-Nummer: 1312-43-2
EINECS: 215-193-9
Merck Index: 14,4952

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR