Tetrachlorgold(III)säure Trihydrat ≥99,99% (Metall-Basis) Au ≥ 49.0% ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Hydrogentetrachloroaurat(III) Trihydrat, Tetrachlorgoldsäure Trihydrat
36400.06
AB208060.0001
36400.06EA
816
EUR
36400.06
ABCRAB208060.0001
Tetrachlorgold(III)säure Trihydrat ≥99,99% (Metall-Basis) Au ≥ 49.0% ACS
Tetrachlorgold(III)säure Trihydrat
Purity: 99.99%
CAS No.: 16961-25-4
Molecular Formula: HAuCl4.3H2O
Formula Weight: 393.83 (339.79anhy)
Physical Form: Crystalline
Sensitivity: Light Sensitive and Hygroscopic
Appearance: Yellow
Odour: Odourless
UN No.: 3260
MDL No.: MFCD00149904
Formel:
HAuCl₄·3H₂O Molecular Weight: 393.83 g/mol Schmelzpunkt: 30 Dichte: 3.9 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149904 CAS-Nummer: 16961-25-4 EINECS: 240-948-4 UN: 3260 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...