L(+)-Weinsäure ACS

Lieferant: Thermo Fisher Scientific
Gefahr

36405.22 36405.36 36405.A3
36405.22EA 44 EUR
36405.22 36405.36 36405.A3
L(+)-Weinsäure ACS
L(+)-Weinsäure

CAS No.: 87-69-4 Molecular Formula: C4H6O6 Formula Weight: 150.09 Physical Form: Granular Density: 1.760 Odour: Odourless pH: 2.1 Melting Point: 168-170° Solubility: Soluble in water, methanol, ethanol, glycerol Beilstein Reference: 1725147 Merck Reference: 14,9070 MDL No.: MFCD00064207


Useful as a resolving agent

Formel: C₄H₆O₆
Siedepunkt: 399 °C (1013 hPa)
Schmelzpunkt: 168…170 °C
Dichte: 1,760
Lagertemperatur: Raumtemperatur
MDL: MFCD00064207
CAS-Nummer: 87-69-4
EINECS: 201-766-0
REACH: 01-2119537204-47
Merck Index: 14,9070
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4e859e52081482a351d52322717614aad450742&vid=879c292903ddee7a59d5d6b2d969c4c8fc6eac75&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR