L(+)-Weinsäure ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
CAS No.: 87-69-4
Molecular Formula: C4H6O6
Formula Weight: 150.09
Physical Form: Granular
Density: 1.760
Odour: Odourless
pH: 2.1
Melting Point: 168-170°
Solubility: Soluble in water, methanol, ethanol, glycerol
Beilstein Reference: 1725147
Merck Reference: 14,9070
MDL No.: MFCD00064207
Useful as a resolving agent
Formel:
C₄H₆O₆ Siedepunkt: 399 °C (1013 hPa) Schmelzpunkt: 168…170 °C Dichte: 1,760 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064207 CAS-Nummer: 87-69-4 EINECS: 201-766-0 REACH: 01-2119537204-47 Merck Index: 14,9070 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...