Cobalt(II)nitrat Hexahydrat 98,0-102,0% ACS

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Cobaltnitrat Hexahydrat, Cobaltdinitrat Hexahydrat

36418.22 36418.36 36418.A4
36418.22EA 80.2 EUR
36418.22 36418.36 36418.A4
Cobalt(II)nitrat Hexahydrat 98,0-102,0% ACS
Cobalt(II)nitrat Hexahydrat

Applications: As a battery additive, in the manufacture of cobalt pigments and invisible inks, for decorating stoneware and procelain, and in the preparation of catalysts
Very soluble in water, alcohol, and most organic solvents


Purity: 98.0-102.0% CAS No.: 10026-22-9 Molecular Formula: Co(NO3)2.6H2O Formula Weight: 291.04 (182.94anhy) Physical Form: Crystalline Density: 1.87 Refractive Index: 1.52 Appearance: Red Odour: Odourless Melting Point: 55-56° Solubility: Very soluble in water, alcohol, and most organic solvents Merck Reference: 14,2444 UN No.: 1477 MDL No.: MFCD00149647


As a battery additive, in the manufacture of cobalt pigments and invisible inks, for decorating stoneware and procelain, and in the preparation of catalysts

Formel: Co(NO₃)₂·6H₂O
Schmelzpunkt: 55…56 °C
Dichte: 1,87
Lagertemperatur: Raumtemperatur
MDL: MFCD00149647
CAS-Nummer: 10026-22-9
EINECS: 233-402-1
UN: 1477
ADR: 5.1,III
Merck Index: 14,2444

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR