Natriumformiat ≥99,0% ACS
Lieferant: Thermo Fisher Scientific
Purity: 99.0% min
CAS No.: 141-53-7
Molecular Formula: HCO2Na
Formula Weight: 68.01
Physical Form: Crystalline
Density: 1.92
Melting Point: 259-262°
Solubility: Soluble in water, glycerol. Slightly soluble in alcohol
Beilstein Reference: 3595134
Merck Reference: 14,8621
MDL No.: MFCD00013101
Reducing agent, buffering agent, complexing agent, Noble metal precipitant
Formel:
HCO₂Na Schmelzpunkt: 259…262 °C Dichte: 1,92 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013101 CAS-Nummer: 141-53-7 EINECS: 205-488-0 REACH: 01-2119486468-21 Merck Index: 14,8621 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...