Nitrite nitrogen standard for IC, certified reference material, TraceCERT®, Supelco®
Lieferant: Merck
36427-100ML
36427-100MLEA 56.1 EUR
36427-100ML
Nitrite nitrogen standard for IC, certified reference material, TraceCERT®, Supelco®
Standards Anionische Standards Nitrit (N)-Standards
Standards: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.