tri-Natriumcitrat Dihydrat 99,0-105,0% (durch Trockensubstanz), körnig USP, Multikompendial, J.T.Baker®

Lieferant: Avantor

3647-01EA 94.39 EUR
3647-01 3647-05 3647-07 3647-20 3647-35 3647-37
tri-Natriumcitrat Dihydrat 99,0-105,0% (durch Trockensubstanz), körnig USP, Multikompendial, J.T.Baker®
tri-Natriumcitrat Dihydrat

  • GMP manufactured product

Formel: Na₃C₆H₅O₇·2H₂O
Schmelzpunkt: 150 °C
Dichte: 1,70 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00150031
CAS-Nummer: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product Passes Test
Meets E.P. Chemical Specifications Passes Test
Meets B.P. Chemical Specifications Passes Test
Meets U.S.P Requirements Passes Test
Meets J.P. Chemical Specifications 10.0 - 13.0%
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical 99.0 - 100.5%
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C Passes Test
USP - Alkalinity Passes Test
USP - Water (H₂O) 10.0 - 13.0 %
USP - Tartrate Passes Test
USP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis) 99.0 - 100.5 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Readily Carbonizable Substances Passes Test
EP/BP - Chloride (Cl) ≤50 ppm
EP/BP - Oxalate (C₂O₄) ≤300 ppm
EP/BP - Sulfate (SO₄) ≤150 ppm
EP/BP - Water (H₂O) 11.0 - 13.0 %
EP/BP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis) 99.0 - 101.0 %
EP/BP - Endotoxin Concentration (EU/g) ≤2.5
JP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis) 99.0 - 101.0 %
JP - pH (1 in 20) 7.5 - 8.5
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤0.015 %
JP - Identification Passes Test
JP - Sulfate (SO₄) ≤0.048 %
JP - Heavy Metals (as Pb) ≤10 ppm
JP - Arsenic (As) ≤2 ppm
JP - Tartrate Passes Test
JP - Oxalate (C₂O₄) Passes Test
JP - Readily Carbonizable Substances Passes Test
JP - Loss on Drying at 180°C 10.0 - 13.0 %
Total Aerobic Microbial Count, For Information Only
Total Yeasts and Molds Count, For Information Only
E. coli Negative
P. aureginosa Negative
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR