tri-Natriumcitrat Dihydrat 99,0-105,0% (durch Trockensubstanz), körnig USP, Multikompendial, J.T.Baker®
Lieferant: Avantor
3647-01EA
94.39
EUR
3647-01
3647-05
3647-07
3647-20
3647-35
3647-37
tri-Natriumcitrat Dihydrat 99,0-105,0% (durch Trockensubstanz), körnig USP, Multikompendial, J.T.Baker®
tri-Natriumcitrat Dihydrat
- GMP manufactured product
Formel:
Na₃C₆H₅O₇·2H₂O Schmelzpunkt: 150 °C Dichte: 1,70 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150031 CAS-Nummer: 6132-04-3 EINECS: 200-675-3 Merck Index: 13,08675 |
|
Spezifikation Testergebnisse
GMP Manufactured Product | Passes Test |
Meets E.P. Chemical Specifications | Passes Test |
Meets B.P. Chemical Specifications | Passes Test |
Meets U.S.P Requirements | Passes Test |
Meets J.P. Chemical Specifications | 10.0 - 13.0% |
CAUTION: For Manufacturing, processing or repackaging | Passes Test |
Bulk Pharmaceutical Chemical | 99.0 - 100.5% |
USP - Identification A | Passes Test |
USP - Identification B | Passes Test |
USP - Identification C | Passes Test |
USP - Alkalinity | Passes Test |
USP - Water (H₂O) | 10.0 - 13.0 % |
USP - Tartrate | Passes Test |
USP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis) | 99.0 - 100.5 % |
EP/BP - Identification A | Passes Test |
EP/BP - Identification B | Passes Test |
EP/BP - Appearance of Solution | Passes Test |
EP/BP - Acidity or Alkalinity | Passes Test |
EP/BP - Readily Carbonizable Substances | Passes Test |
EP/BP - Chloride (Cl) | ≤50 ppm |
EP/BP - Oxalate (C₂O₄) | ≤300 ppm |
EP/BP - Sulfate (SO₄) | ≤150 ppm |
EP/BP - Water (H₂O) | 11.0 - 13.0 % |
EP/BP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis) | 99.0 - 101.0 % |
EP/BP - Endotoxin Concentration (EU/g) | ≤2.5 |
JP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis) | 99.0 - 101.0 % |
JP - pH (1 in 20) | 7.5 - 8.5 |
JP - Clarity and Color of Solution | Passes Test |
JP - Chloride (Cl) | ≤0.015 % |
JP - Identification | Passes Test |
JP - Sulfate (SO₄) | ≤0.048 % |
JP - Heavy Metals (as Pb) | ≤10 ppm |
JP - Arsenic (As) | ≤2 ppm |
JP - Tartrate | Passes Test |
JP - Oxalate (C₂O₄) | Passes Test |
JP - Readily Carbonizable Substances | Passes Test |
JP - Loss on Drying at 180°C | 10.0 - 13.0 % |
Total Aerobic Microbial Count, For Information Only | |
Total Yeasts and Molds Count, For Information Only | |
E. coli | Negative |
P. aureginosa | Negative |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...