Niob(V)oxid ≥99,95% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

36478.14 36478.22
36478.14EA 107 EUR
36478.14 36478.22
Niob(V)oxid ≥99,95% (Metall-Basis)
Niob(V)oxid

Purity: 99.95% CAS No.: 1313-96-8 Molecular Formula: Nb2O5 Formula Weight: 265.81 Physical Form: 3-12mm Sintered Lump Density: 4.6 Melting Point: 1520° Merck Reference: 14,6559 MDL No.: MFCD00011128


For vacuum deposition

Formel: Nb₂O₅
Schmelzpunkt: 1520 °C
Dichte: 4,6
Lagertemperatur: Raumtemperatur
MDL: MFCD00011128
CAS-Nummer: 1313-96-8
EINECS: 215-213-6
Merck Index: 14,6559

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR