Iodwasserstoffsäure 55 - 58% ACS

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Hydrogeniodid

36484.AD 36484.AP 36484.AY
36484.ADEA 62.3 EUR
36484.AD 36484.AP 36484.AY
Iodwasserstoffsäure 55 - 58% ACS
Hydriodic acid

Suitable for use in methoxyl determinations.


Used as catalyst for the conversion of primary alcohol to alkyl halide.


Bitte beachten Light sensitive liquid.

Formel: HI
Molecular Weight: 127.91 g/mol
Dichte: 1.701
Lagertemperatur: Raumtemperatur
MDL: MFCD00011347
CAS-Nummer: 10034-85-2
EINECS: 233-109-9
UN: 1787
ADR: 8 II
Merck Index: 14,4776

Bestellen

Spezifikation Testergebnisse

Assay (Unspecified) 55.0 - 58.0%
Residue on Ignition Max. 0.01%
Impurity Content, Chloride and Bromide (as Cl) Max. 0.05%
Impurity Content, Fe Max. 0.001%
Impurity Content, I₂ Max. 0.75%
Impurity Content, PO₄ Max. 0.001%
Impurity Content, SO₄ Max. 0.005%
Trace Metals (as Pb) Max. 0.001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR