D(+)-Saccharose ACS
Lieferant: Thermo Fisher Scientific
Synonyme:
Haushaltszucker, D(+)-Sucrose, Rübenzucker, Rohrzucker, 2-β-D-Fructofuranosyl-1-α-D-glucopyranosid, Sucrose, Kristallzucker
36508.30
36508.A1
36508.30EA
42.8
EUR
36508.30
36508.A1
D(+)-Saccharose ACS
D(+)-Saccharose
CAS No.: 57-50-1
Molecular Formula: C12H22O11
Formula Weight: 342.30
Physical Form: Crystalline
Density: 1.58
Appearance: White
Odour: Odourless
Melting Point: 185-187°
Solubility: Soluble in water. Slightly soluble in alcohol, methanol
Beilstein Reference: 90825
Merck Reference: 14,8881
MDL No.: MFCD00006626
Formel:
C₁₂H₂₂O₁₁ Molecular Weight: 342.3 g/mol Schmelzpunkt: 185…187 °C Dichte: 1.58 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006626 CAS-Nummer: 57-50-1 EINECS: 200-334-9 Merck Index: 14,8881 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...