Mangan(II)chlorid Tetrahydrat 98,0-101,0% ACS

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Mangandichlorid Tetrahydrat

36526.22 36526.36 36526.A3
36526.22EA 33 EUR
36526.22 36526.36 36526.A3
Mangan(II)chlorid Tetrahydrat 98,0-101,0% ACS
Mangan(II)chlorid Tetrahydrat

Purity: 98.0-101.0% CAS No.: 13446-34-9 Molecular Formula: Cl2Mn.4H2O Formula Weight: 197.90 (125.84anhy) Physical Form: Crystalline Density: 2.010 Appearance: Pink Odour: Odourless Melting Point: 58° Solubility: Soluble in water, alcohol Merck Reference: 14,5728 MDL No.: MFCD00149792

Formel: MnCl₂·4H₂O
Siedepunkt: 1190 °C (1013 hPa)
Schmelzpunkt: 58 °C
Dichte: 2,010
Lagertemperatur: Raumtemperatur
MDL: MFCD00149792
CAS-Nummer: 13446-34-9
EINECS: 231-869-6
UN: 3077
ADR: 9,III
Merck Index: 14,5728

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR