HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat) 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
HATU, 1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxide, hexafluorophosphate
365310050
365310250
365310050.EA
117
EUR
365310050.
365310250.
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat) 97%
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat)
Formel:
C₁₀H₁₅F₆N₆OP Schmelzpunkt: 189…191 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00274639 CAS-Nummer: 148893-10-1 UN: 1325 ADR: 4.1,II |
Spezifikation Testergebnisse
Appearance | White to beige Crystalline powder and/or lumps |
Infrared spectrum | Conforms |
Melting point | 183 to 193 °C (Dec) |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...