Pararosaniliniumchlorid ≥90%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Basic Red 9, Fuchsin basisch Hydrochlorid, 4,4'-(4-Iminocyclohexa-2,5-dienylidenmethylen)dianilinhydrochlorid, 4,4',4''-Triaminotrityliumchlorid, 4,4′-(4-Iminocyclohexa-2,5-dienylidenmethylen)dianilinhydrochlorid, Pararosanilin, Parafuchsin
36540.18
36540.18EA
123
EUR
36540.18
Pararosaniliniumchlorid ≥90%
Pararosaniliniumchlorid
Purity: 90+%
CAS No.: 569-61-9
Molecular Formula: C19H17N3.HCl
Formula Weight: 323.83
Physical Form: Powder
Melting Point: 268-270° dec.
Beilstein Reference: 4164603
MDL No.: MFCD00001657
pH indicator 1.2 (purple) to 3.0 (red)
Formel:
C₁₉H₁₈ClN₃ Molecular Weight: 323,82 g/mol Schmelzpunkt: 268…270 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001657 CAS-Nummer: 569-61-9 EINECS: 209-321-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...