Tin(IV) isopropoxide 10% (w/v) in 2-Propanol/ Toluol , 98% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

36563.AA 36563.AE
36563.AAEA 41.2 EUR
36563.AA 36563.AE
Tin(IV) isopropoxide 10% (w/v) in 2-Propanol/ Toluol , 98% (Metall-Basis)
Tin(IV) isopropoxide

Purity: 98% CAS No.: 1184-61-8 Molecular Formula: Sn[OCH(CH3)2]4 Formula Weight: 355.06 Physical Form: Liquid Sensitivity: Moisture Sensitive UN No.: UN1993 MDL No.: MFCD00145408

Formel: C₁₂H₂₈O₄Sn
Molecular Weight: 355.062 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00145408
CAS-Nummer: 1184-61-8
UN: 1993
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR