Neodymium(III) isopropoxide 2,5% (w/v) in 2-Propanol/ Toluol , 99% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Neodymium(III) i-propoxide
36573.AC
36573.AE
36573.ACEA
124
EUR
36573.AC
36573.AE
Neodymium(III) isopropoxide 2,5% (w/v) in 2-Propanol/ Toluol , 99% (REO, Basis der Oxide der Metalle der seltenen Erden)
Neodymium(III) isopropoxide
Purity: 99%
CAS No.: 19236-15-8
Molecular Formula: C9H21NdO3
Formula Weight: 321.30
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Appearance: Blue
UN No.: UN1993
MDL No.: MFCD00058819
Formel:
C₉H₂₁NdO₃ Molecular Weight: 321.503 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00058819 CAS-Nummer: 19236-15-8 UN: 1993 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...