Aluminum calcium isopropoxide 10% (w/v) in 2-Propanol (IPA) , 99.8% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99.8%
CAS No.: 23275-27-6
Molecular Formula: Ca[Al(OCH(CH3)2)4]2
Formula Weight: 566.30
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Odour: Alcohol-like
Boiling Point: 230-240°/4.0mm
UN No.: UN1219
MDL No.: MFCD00143586
Formel:
Ca[Al(OCH(CH₃)₂)₄]₂ Siedepunkt: 230…240 °C (4,0 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143586 CAS-Nummer: 23275-27-6 UN: 1219 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...