Thioharnstoff ≥99% ACS

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Aminomercaptoameisensäureamid, Thiocarbamid

36609.18 36609.30 36609.A1
36609.18EA 42.5 EUR
36609.18 36609.30 36609.A1
Thioharnstoff ≥99% ACS
Thioharnstoff

Purity: 99% min. CAS No.: 62-56-6 Molecular Formula: CH4N2S Formula Weight: 76.12 Physical Form: Crystalline Density: 1.405 Appearance: White Odour: Odourless Melting Point: 175-178° Beilstein Reference: 605327 Merck Reference: 14,9367 UN No.: 3077 MDL No.: MFCD00008067


Reagent for organic synthesis

Formel: NH₂CSNH₂
Molecular Weight: 76.12 g/mol
Schmelzpunkt: 175…178 °C
Dichte: 1.405
Lagertemperatur: Raumtemperatur
MDL: MFCD00008067
CAS-Nummer: 62-56-6
EINECS: 200-543-5
UN: 3077
ADR: 9 III
Merck Index: 14,9367
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fff6222c54fe16ea4d36edcbf320745cb1e18d6&vid=e9ae5dc49e92d68f893244a5cd4e0245f289ed09&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR