Thioharnstoff ≥99% ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Aminomercaptoameisensäureamid, Thiocarbamid
36609.18
36609.30
36609.A1
36609.18EA
42.5
EUR
36609.18
36609.30
36609.A1
Thioharnstoff ≥99% ACS
Thioharnstoff
Purity: 99% min.
CAS No.: 62-56-6
Molecular Formula: CH4N2S
Formula Weight: 76.12
Physical Form: Crystalline
Density: 1.405
Appearance: White
Odour: Odourless
Melting Point: 175-178°
Beilstein Reference: 605327
Merck Reference: 14,9367
UN No.: 3077
MDL No.: MFCD00008067
Reagent for organic synthesis
Formel:
NH₂CSNH₂ Molecular Weight: 76.12 g/mol Schmelzpunkt: 175…178 °C Dichte: 1.405 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008067 CAS-Nummer: 62-56-6 EINECS: 200-543-5 UN: 3077 ADR: 9 III Merck Index: 14,9367 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...