Diphenylamin ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
CAS No.: 122-39-4
Molecular Formula: C12H11N
Formula Weight: 169.23
Flash Point: 152°(305°F)
Physical Form: Solid
Density: 1.16
Appearance: White to off-white
Melting Point: 52-55°
Boiling Point: 302°
Solubility: Soluble in benzene, ether, glacial acetic acid, CS2
Beilstein Reference: 508755
Merck Reference: 14,3317
UN No.: 3077
MDL No.: MFCD00003014
Formel:
(C₆H₅)₂NH Siedepunkt: 302 °C Schmelzpunkt: 52…55 °C Dichte: 1,16 Flammpunkt: 152 °C (305 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003014 CAS-Nummer: 122-39-4 EINECS: 204-539-4 UN: 3077 ADR: 9,III Merck Index: 14,3317 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...