Erbium(III)isopropoxid

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Erbium(III) i-propoxide

36613.03 36613.06
36613.03EA 99.5 EUR
36613.03 36613.06
Erbium(III)isopropoxid
Erbium(III)isopropoxid

CAS No.: 14814-07-4 Molecular Formula: C9H21ErO3 Formula Weight: 344.52 Flash Point: 58°(138°F) Physical Form: Powder Sensitivity: Air and Moisture Sensitive UN No.: 2925 MDL No.: MFCD00144253

Formel: C₉H₂₁ErO₃
Molecular Weight: 344.52 g/mol
Schmelzpunkt: 325 °C (Zers.)
Flammpunkt: 58 °C (138 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00144253
CAS-Nummer: 14814-07-4
ADR: 4.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR