Erbium(III)isopropoxid
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Erbium(III) i-propoxide
36613.03
36613.06
36613.03EA
99.5
EUR
36613.03
36613.06
Erbium(III)isopropoxid
Erbium(III)isopropoxid
CAS No.: 14814-07-4
Molecular Formula: C9H21ErO3
Formula Weight: 344.52
Flash Point: 58°(138°F)
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
UN No.: 2925
MDL No.: MFCD00144253
Formel:
C₉H₂₁ErO₃ Molecular Weight: 344.52 g/mol Schmelzpunkt: 325 °C (Zers.) Flammpunkt: 58 °C (138 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00144253 CAS-Nummer: 14814-07-4 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...