Glycerin ≥99,5% ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1,2,3-Propantriol, Glycerol
36646.AP
36646.K2
36646.M1
36646.APEA
60.4
EUR
36646.AP
36646.K2
36646.M1
Glycerin ≥99,5% ACS
Glycerin
Purity: 99.5+%
CAS No.: 56-81-5
Molecular Formula: C3H8O3
Formula Weight: 92.09
Flash Point: 160°(320°F)
Physical Form: Viscous liquid
Density: 1.26
Refractive Index: 1.474
Sensitivity: Hygroscopic
Appearance: Colourless
Odour: Odourless
Melting Point: 18°
Boiling Point: 182°/20mm
Beilstein Reference: 635685
Merck Reference: 14,4484
MDL No.: MFCD00004722
Formel:
HOCH₂CH(OH)CH₂OH Siedepunkt: 182 °C (20 mmHg) Schmelzpunkt: 18 °C Dichte: 1,26 Flammpunkt: 160 °C (320 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004722 CAS-Nummer: 56-81-5 EINECS: 200-289-5 Merck Index: 14,4484 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...