Yttrium(III)-2-ethylhexanoat ≥99,8% (Metall-Basis)

Lieferant: Thermo Scientific
Achtung

36661.14
36661.14EA 168 EUR
36661.14
Yttrium(III)-2-ethylhexanoat ≥99,8% (Metall-Basis)
Yttrium(III)-2-ethylhexanoat

Purity: 99.8% CAS No.: 103470-68-4 Molecular Formula: C24H45O6Y Formula Weight: 518.16 Physical Form: Powder

Formel: C₂₄H₄₅O₆Y
Molecular Weight: 518,524 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00070441
CAS-Nummer: 103470-68-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR