Titan ≥99,999% (Metall-Basis), Teller, Puratronic® 100x100 mm (3.9x3.9 in), Dicke 6 mm (0.2 in)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
36680.KG
36680.KGEA
739
EUR
36680.KG
Titan ≥99,999% (Metall-Basis), Teller, Puratronic® 100x100 mm (3.9x3.9 in), Dicke 6 mm (0.2 in)
Titanium
Purity: 99.999%
CAS No.: 7440-32-6
Physical Form: ≈271g/pc
Odour: Odourless
MDL No.: MFCD00011264
Formel:
Ti Molecular Weight: 47.88 g/mol Siedepunkt: 3262 Schmelzpunkt: 1660 Dichte: 4.51 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011264 CAS-Nummer: 7440-32-6 EINECS: 231-142-3 Merck Index: 12,09610 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...