Natriumdichromat Dihydrat 99,5-100,5% ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
36692.22
36692.36
36692.A3
36692.PG
36692.22EA
40.7
EUR
36692.22
36692.36
36692.A3
36692.PG
Natriumdichromat Dihydrat 99,5-100,5% ACS
Natriumdichromat Dihydrat
Purity: 99.5-100.5%
CAS No.: 7789-12-0
Molecular Formula: Cr2Na2O7.2H2O
Formula Weight: 298.00 (261.97anhy)
Physical Form: Crystalline
Density: 2.348
Appearance: Red
Odour: Odourless
Melting Point: 100° -2H2O
Merck Reference: 14,8609
UN No.: 3087
MDL No.: MFCD00149166
Formel:
Na₂Cr₂O₇·2H₂O Molecular Weight: 298.00 g/mol (261.97 anhy) Schmelzpunkt: 100 °C (-2H₂O) Dichte: 2.348 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149166 CAS-Nummer: 7789-12-0 EINECS: 234-190-3 UN: 3087 ADR: 5.1 II Merck Index: 14,8609 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...