Bromwasserstoff 47,0 - 49,0% ACS

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Bromwasserstoff, Hydrogenbromid

36694.AK 36694.K2 36694.M1
36694.AKEA 58.4 EUR
36694.AK 36694.K2 36694.M1
Bromwasserstoff 47,0 - 49,0% ACS
Bromwasserstoff

CAS No.: 10035-10-6 Molecular Formula: HBr Formula Weight: 80.92 Physical Form: Liquid Density: 1.490 Sensitivity: Air Sensitive Appearance: Colourless to yellow Beilstein Reference: 3587158 Merck Reference: 14,4778 UN No.: 1788 MDL No.: MFCD00011323

Formel: HBr
Molecular Weight: 80.91 g/mol
Siedepunkt: -66.7
Schmelzpunkt: -86
Dichte: 1.490
Lagertemperatur: Raumtemperatur
MDL: MFCD00011323
CAS-Nummer: 10035-10-6
EINECS: 233-113-0
UN: 1788
ADR: 8 II
Merck Index: 14,4778

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR