Stärke, löslich ACS Iodometric Titrations
Lieferant: Thermo Fisher Scientific
Synonyme:
Amylodextrin, Vitex® Iodindikator
36703.18
36703.30
36703.A1
36703.18EA
37.2
EUR
36703.18
36703.30
36703.A1
Stärke, löslich ACS Iodometric Titrations
Stärke, löslich
CAS No.: 9005-84-9
Physical Form: Powder
Odour: Odourless
Melting Point: 256-258° dec.
Solubility: Soluble in hot water
Merck Reference: 14,8799
MDL No.: MFCD00082026
Formel:
(C₆H₁₀O₅)n Molecular Weight: 162.14 g/mol Schmelzpunkt: 256…258 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082026 CAS-Nummer: 9005-84-9 EINECS: 232-686-4 Merck Index: 14,8799 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...