Platin ≥99,9% (Metall-Basis), amorphes Pulver APS <3 µm

Lieferant: Thermo Fisher Scientific

Synonyme: Platinium

36704.03 36704.06
36704.03EA 278 EUR
36704.03 36704.06
Platin ≥99,9% (Metall-Basis), amorphes Pulver APS <3 µm
Platinum

Purity: 99.9% CAS No.: 7440-06-4 Physical Form: S.A. 3.5-7.9m2/g, Tap density 0.6-2.0g/cm3 Odour: Odourless MDL No.: MFCD00011179

Formel: Pt
Molecular Weight: 195.08 g/mol
Siedepunkt: 3827
Schmelzpunkt: 1769
Dichte: 21.4 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011179
CAS-Nummer: 7440-06-4
EINECS: 231-116-1
UN: 3089
ADR: 4.1 II
Merck Index: 12,07689

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR