Nitroethane-1,1-d₂ (99% D), purum, Supelco®

Lieferant: Merck

36766-250MG-R
36766-250MG-REA 47.9 EUR
36766-250MG-R
Nitroethane-1,1-d₂ (99% D), purum, Supelco®
Nitroethane-1,1-d₂
Formel: CH₃CD₂NO₂
Molecular Weight: 77,08 g/mol
MDL: MFCD00192352
CAS-Nummer: 13031-33-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR