Oxalyldichlorid 2.0 M in Dichlormethan, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

369181000 369188000
369181000.EA 50.6 EUR
369181000. 369188000.
Oxalyldichlorid 2.0 M in Dichlormethan, AcroSeal®
Oxalyldichlorid
Formel: C₂Cl₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD00000704
CAS-Nummer: 79-37-8
EINECS: 201-200-2
UN: 2922
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Solution
Titration Argentometric 1.9 to 2.1 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR