Hemin chloride (aus Schwein), Calbiochem®, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: Chlorohemin, 1,3,5,8-Tetramethyl-2,4-divinylporphine-6,7-dipropionic acid ferrichloride, Chloroprotoporphyrin IX iron(III), Haemin, Hemin, Ferriprotoporphyrin IX chloride

M-Clarity™ Qualitätsstufe = MQ100

3741-5
3741-5EA 149 EUR
3741-5
Hemin chloride (aus Schwein), Calbiochem®, Millipore®
Hemin chloride

Iron-containing compound of porcine origin that can induce hydrogen peroxide-dependent lipid peroxidation. Reported to increase the number and size of myotubes and the frequency of myotube contractions.

Formel: C₃₄H₃₂ClFeN₄O₄
Molecular Weight: 651,95 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010726
CAS-Nummer: 16009-13-5
EINECS: 240-140-1
Merck Index: 14,04644

Bestellen

Spezifikation Testergebnisse

Purity Homogeneous by TLC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR