Bovine haemoglobin (from Erythrocytes)
Lieferant: Merck Millipore Calbiochem
374834-10
374834-10EA
101
EUR
374834-10
Bovine haemoglobin (from Erythrocytes)
Proteine und Peptide
Hemoglobin, bovine erythrocytes is an iron-containing oxygen-transport metalloprotein. Suitable as a substrate for proteases, such as papain, pepsin, and trypsin.
- Lyophilised form
- Standard Handling
- Glass bottles used for packaging