Biotinylated heparin, porcine intestinal mucosa, Calbiochem®
Lieferant: Merck Millipore Calbiochem
375054-10
375054-10EA 288 EUR
375054-10
Biotinylated heparin, porcine intestinal mucosa, Calbiochem®
Proteinkonjugate und Markierungen
Biotinylated heparin contains 5 − 8 mole-percent biotin. Biotin attachment is through a 12-atom spacer arm containing a cleavable disulfide group.