1,4-Dimethylpyridinium iodide 99%, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
Synonyme:
1,4-Dimethylpyridin-1-ium iodide
376434-25G
376434-25GEA
130
EUR
376434-25G
1,4-Dimethylpyridinium iodide 99%, Sigma-Aldrich®
1,4-Dimethylpyridinium iodide
1,4-Dimethylpyridinium iodide,99%
Formel:
C₇HIN Molecular Weight: 235.07 g/mol Schmelzpunkt: 151…154 °C (Lit.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00031899 CAS-Nummer: 2301-80-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...