Titan(IV)chlorid 1 M in Dichlormethan, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

377741000 377748000
377741000.EA 97.6 EUR
377741000. 377748000.
Titan(IV)chlorid 1 M in Dichlormethan, AcroSeal®
Titan(IV)chlorid
Formel: Cl₄Ti
Lagertemperatur: Raumtemperatur
MDL: MFCD00011267
CAS-Nummer: 7550-45-0
EINECS: 231-441-9
UN: 2922
ADR: 6.1,II

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow to dark brown Solution
Titration Argentometric 0.95 to 1.10 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR