1-Heptyl-4-(4-pyridyl)pyridinium bromide ≥95%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: 1-n-Heptyl-4-(4-pyridyl)pyridinium bromide

377783-5G
377783-5GEA 278 EUR
377783-5G
1-Heptyl-4-(4-pyridyl)pyridinium bromide ≥95%, Sigma-Aldrich®
1-Heptyl-4-(4-pyridyl)pyridinium bromide

1-Heptyl-4-(4-pyridyl)pyridinium bromide,95%

Formel: C₁₇H₂₃BrN₂
Molecular Weight: 335,289 g/mol
Schmelzpunkt: 125…128 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00134461
CAS-Nummer: 39127-10-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ec1d40dbbdafe4f20d7c213154e0e2ffed8e798&vid=905d367c7f5d40b30d5e30cead901587d614b794&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR